accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOOOCH3

CAS RN 57153-18-1

IUPAC Name: methyl 4-(4-chloro-2-methylphenoxy)butanoate
Synonyms: methyl 4-(4-chloro-2-methylphenoxy)butanoate
Molecular Weight (g/mol): 242.7
Molecular Formula: C12H15ClO3
InChi Key: FWDQLSHRVKQKBS-UHFFFAOYSA-N
SMILES: COC(=O)CCCOC1=C(C)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results