accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57153-43-2

IUPAC Name: 2-{4-methyl-4H,6H,7H-thieno[3,2-c]pyran-4-yl}acetic acid
Synonyms: {4-methyl-6H,7H-thieno[3,2-c]pyran-4-yl}acetic acid
Molecular Weight (g/mol): 212.26
Molecular Formula: C10H12O3S
InChi Key: MBZWBAMMUGJINB-UHFFFAOYNA-N
SMILES: CC1(CC(O)=O)OCCC2=C1C=CS2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results