accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH

CAS RN 57184-23-3

IUPAC Name: 1-(cyclohexylmethyl)piperazine
Synonyms: 1-(cyclohexylmethyl)piperazine
Molecular Weight (g/mol): 182.31
Molecular Formula: C11H22N2
InChi Key: LRPGNFROBDUREU-UHFFFAOYSA-N
SMILES: C(C1CCCCC1)N1CCNCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)