accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57238-75-2

IUPAC Name: 5-(chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
Synonyms: 5-(chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 229.06
Molecular Formula: C9H6Cl2N2O
InChi Key: BJVYSQGEJHKTBW-UHFFFAOYSA-N
SMILES: ClCC1=NC(=NO1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)