accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONN

CAS RN 57260-70-5

IUPAC Name: tert-butyl 4-benzylpiperazine-1-carboxylate
Synonyms: tert-butyl 4-benzylpiperazine-1-carboxylate
Molecular Weight (g/mol): 276.38
Molecular Formula: C16H24N2O2
InChi Key: GVHSMUYEAWMYLM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC=CC=C2)CC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results