accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNSNCl

CAS RN 5728-20-1

IUPAC Name: dichloro-1,2,5-thiadiazole
Synonyms: dichloro-1,2,5-thiadiazole
Molecular Weight (g/mol): 155
Molecular Formula: C2Cl2N2S
InChi Key: YNZQOVYRCBAMEU-UHFFFAOYSA-N
SMILES: ClC1=NSN=C1Cl
Boiling Point 
  • (3)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)