accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57292-44-1

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-chlorophenyl)propanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(4-chlorophenyl)propanoic acid| BOC-4-CHLORO-D-PHENYLALANINE
Molecular Weight (g/mol): 299.75
Molecular Formula: C14H18ClNO4
InChi Key: BETBOAZCLSJOBQ-LLVKDONJSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results