accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONH(R)FOO

CAS RN 57292-45-2

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(4-fluorophenyl)propanoate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H17FNO4
InChi Key: RCXSXRAUMLKRRL-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(F)C=C1)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results