accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57294-38-9

IUPAC Name: 4-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: 4-[(tert-butoxycarbonyl)amino]butanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: HIDJWBGOQFTDLU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCC(O)=O
Quantity 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)