accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 573-17-1

IUPAC Name: 9-bromophenanthrene
Synonyms: 9-bromophenanthrene
Molecular Weight (g/mol): 257.13
Molecular Formula: C14H9Br
InChi Key: RSQXKVWKJVUZDG-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=CC=C2C2=CC=CC=C12
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)