accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OO

CAS RN 57319-65-0

IUPAC Name: 6-amino-1,3-dihydro-2-benzofuran-1-one
Synonyms: 6-amino-3H-2-benzofuran-1-one
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: ZIJZDNKZJZUROE-UHFFFAOYSA-N
SMILES: NC1=CC=C2COC(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results