accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOCH2

CAS RN 57322-50-6

IUPAC Name: N-ethyl-2-phenyl-N-[(pyridin-4-yl)methyl]prop-2-enamide
Synonyms: N-ethyl-2-phenyl-N-(pyridin-4-ylmethyl)prop-2-enamide
Molecular Weight (g/mol): 266.34
Molecular Formula: C17H18N2O
InChi Key: LOVSQYAVWGMIRV-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=NC=C1)C(=O)C(=C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results