accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5733-86-8

HOONO

CAS RN 5733-86-8

IUPAC Name: 1-benzyl-5-oxopyrrolidine-3-carboxylic acid
Synonyms: 1-benzyl-5-oxopyrrolidine-3-carboxylic acid
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: VYKQDWPBYULGPF-UHFFFAOYNA-N
SMILES: OC(=O)C1CN(CC2=CC=CC=C2)C(=O)C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results