CAS RN 57344-86-2
IUPAC Name:
3-phenylcyclohexan-1-one
Synonyms:
3-phenylcyclohexan-1-one
Molecular Weight (g/mol):
174.24
Molecular Formula:
C12H14O
InChi Key:
CJAUDSQXFVZPTO-UHFFFAOYNA-N
SMILES:
O=C1CCCC(C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(S)-3-Phenylcyclohexanone 98.0+%, TCI America™
CAS: 57344-86-2 Molecular Formula: C12H14O Molecular Weight (g/mol): 174.24 MDL Number: MFCD08276411 InChI Key: CJAUDSQXFVZPTO-UHFFFAOYNA-N PubChem CID: 11116555 IUPAC Name: 3-phenylcyclohexan-1-one SMILES: O=C1CCCC(C1)C1=CC=CC=C1