accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NON

CAS RN 57351-99-2

IUPAC Name: 5-methyl-1,2-oxazole-3-carbonitrile
Synonyms: 5-methyl-1,2-oxazole-3-carbonitrile
Molecular Weight (g/mol): 108.1
Molecular Formula: C5H4N2O
InChi Key: JTPIMYCMWLOEDD-UHFFFAOYSA-N
SMILES: CC1=CC(=NO1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results