accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3O

CAS RN 5736-86-7

IUPAC Name: 1-(4-propoxyphenyl)ethan-1-one
Synonyms: 1-(4-propoxyphenyl)ethanone
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: RTYYKCQJSTZADZ-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=C1)C(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results