accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrNNOO

CAS RN 573675-25-9

IUPAC Name: 5-bromo-3-nitropyridine-2-carbonitrile
Synonyms: 5-bromo-3-nitropyridine-2-carbonitrile
Molecular Weight (g/mol): 228.01
Molecular Formula: C6H2BrN3O2
InChi Key: OBVYONPVHIOJJZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CN=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results