accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNCl

CAS RN 57381-44-9

IUPAC Name: 5-bromo-2-chlorobenzonitrile
Synonyms: 5-bromo-2-chlorobenzonitrile
Molecular Weight (g/mol): 216.46
Molecular Formula: C7H3BrClN
InChi Key: NKDMQBUJOXQVEA-UHFFFAOYSA-N
SMILES: ClC1=C(C=C(Br)C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results