accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 574-00-5

HOOH

CAS RN 574-00-5

IUPAC Name: naphthalene-1,2-diol
Synonyms: naphthalene-1,2-diol 1,2-naphthohydroquinone β-naphthohydroquinone
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: NXPPAOGUKPJVDI-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CC=CC2=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)