accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 574-19-6

IUPAC Name: 1-(2-hydroxynaphthalen-1-yl)ethan-1-one
Synonyms: 1-(2-hydroxynaphthalen-1-yl)ethanone
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: VUIOUIWZVKVFCI-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2C=CC=CC2=CC=C1O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)