accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57402-36-5

H3CO

CAS RN 57402-36-5

IUPAC Name: docosanal
Synonyms: behenoyl
Molecular Weight (g/mol): 324.59
Molecular Formula: C22H44O
InChi Key: ULCXRAFXRZTNRO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results