accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBOHOH(E)

CAS RN 57404-76-9

IUPAC Name: [(1E)-hept-1-en-1-yl]boronic acid
Synonyms: (1E)-hept-1-en-1-ylboronic acid
Molecular Weight (g/mol): 142.01
Molecular Formula: C7H15BO2
InChi Key: LDTJUGVTOZBIBN-VOTSOKGWSA-N
SMILES: CCCCC\C=C\B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results