accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OCaOH(S)OH(R)OOCHOOOH(S)OH(R)OOCHOO

CAS RN 5743-28-2

IUPAC Name: calcium bis((5R)-5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide) dihydrate
Synonyms: calcium bis((5R)-5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide) dihydrate
Molecular Weight (g/mol): 426.34
Molecular Formula: C12H18CaO14
InChi Key: XBTRVIGNXIDJBU-FWCDDDAWSA-N
SMILES: O.O.[Ca++].OC[C@H](O)[C@H]1OC(=O)[C-](O)C1=O.OC[C@H](O)[C@H]1OC(=O)[C-](O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)