accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57455-06-8

IUPAC Name: (3-iodophenyl)methanol
Synonyms: (3-iodophenyl)methanol
Molecular Weight (g/mol): 234.04
Molecular Formula: C7H7IO
InChi Key: QGCCNWSXJHGUNL-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(I)=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)