accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OOH2COOCH2

CAS RN 57472-68-1

IUPAC Name: 1-[2-(prop-2-enoyloxy)propoxy]propan-2-yl prop-2-enoate
Synonyms: dipropylene glycol diacrylate
Molecular Weight (g/mol): 242.27
Molecular Formula: C12H18O5
InChi Key: RNJVAUBBYGWVBF-UHFFFAOYNA-N
SMILES: CC(COCC(C)OC(=O)C=C)OC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results