accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 575-41-7

IUPAC Name: 1,3-dimethylnaphthalene
Synonyms: 1,3-dimethylnaphthalene 1,3-dimethylnapthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: QHJMFSMPSZREIF-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)