accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 575-44-0

IUPAC Name: naphthalene-1,6-diol
Synonyms: 1,6-dihydroxy naphthalene
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: FZZQNEVOYIYFPF-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(O)=CC=CC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)