accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 575-89-3

IUPAC Name: 2-(2,4,6-trichlorophenoxy)acetic acid
Synonyms: 2,4,6-trichlorophenoxyacetic acid
Molecular Weight (g/mol): 255.48
Molecular Formula: C8H5Cl3O3
InChi Key: KZDCLQBOHGBWOI-UHFFFAOYSA-N
SMILES: OC(=O)COC1=C(Cl)C=C(Cl)C=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results