accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOS

CAS RN 5751-80-4

IUPAC Name: ethyl thiophene-3-carboxylate
Synonyms: ethyl thiophene-3-carboxylate
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: OYSLMAQEMAJMCL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)