accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONOONOHOO

CAS RN 57583-53-6

IUPAC Name: 2,6-dihydroxy-1H,2H,3H,5H,6H,7H-pyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
Synonyms: 2,6-dihydroxypyrrolo[3,4-f]isoindole-1,3,5,7-tetrone
Molecular Weight (g/mol): 248.15
Molecular Formula: C10H4N2O6
InChi Key: VBBUVIWLZBAEEF-UHFFFAOYSA-N
SMILES: ON1C(=O)C2=CC3=C(C=C2C1=O)C(=O)N(O)C3=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results