accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 576-15-8

IUPAC Name: 1-(1H-indol-1-yl)ethan-1-one
Synonyms: 1-(indol-1-yl)ethanone
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: UUCUQJHYUPXDHN-UHFFFAOYSA-N
SMILES: CC(=O)N1C=CC2=CC=CC=C12
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results