accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NOOOONH2

CAS RN 57609-64-0

IUPAC Name: 3-(4-aminobenzoyloxy)propyl 4-aminobenzoate
Synonyms: 3-(4-aminobenzoyloxy)propyl 4-aminobenzoate
Molecular Weight (g/mol): 314.34
Molecular Formula: C17H18N2O4
InChi Key: YPACMOORZSDQDQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(=O)OCCCOC(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results