accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONOO

CAS RN 57620-49-2

IUPAC Name: 4-methyl-4-nitropentanal
Synonyms: 4-methyl-4-nitropentanal
Molecular Weight (g/mol): 145.16
Molecular Formula: C6H11NO3
InChi Key: YDVAFDALOGXWTO-UHFFFAOYSA-N
SMILES: CC(C)(CCC=O)[N+]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results