accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57654-36-1

NNNNNN

CAS RN 57654-36-1

IUPAC Name: bis(pyridin-4-yl)-1,2,4,5-tetrazine
Synonyms: bis(pyridin-4-yl)-1,2,4,5-tetrazine
Molecular Weight (g/mol): 236.24
Molecular Formula: C12H8N6
InChi Key: MQSMIIJCRWDMDL-UHFFFAOYSA-N
SMILES: C1=CC(=CC=N1)C1=NN=C(N=N1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results