accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 57661-23-1

IUPAC Name: hexadecan-6-one
Synonyms: hexadecan-6-one
Molecular Weight (g/mol): 240.43
Molecular Formula: C16H32O
InChi Key: CIAHHJXQZXLMPN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(=O)CCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results