accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57665-05-1

IUPAC Name: sodium pyridine-2-carboxylate
Synonyms: sodium picolinate
Molecular Weight (g/mol): 145.09
Molecular Formula: C6H4NNaO2
InChi Key: RLEBKHAOAHYZHT-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results