accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57671-28-0

IUPAC Name: 1-phenyl-2,5,8,11,14-pentaoxahexadecan-16-ol
Synonyms: 1-phenyl-2,5,8,11,14-pentaoxahexadecan-16-ol
Molecular Weight (g/mol): 328.41
Molecular Formula: C17H28O6
InChi Key: UMUSOTNGYAALST-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results