accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPCH3H3CCH3Br

CAS RN 57702-65-5

IUPAC Name: tributyl(octyl)phosphanium bromide
Synonyms: tributyl(octyl)phosphanium bromide
Molecular Weight (g/mol): 395.45
Molecular Formula: C20H44BrP
InChi Key: UJMLRSWRUXXZEW-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[P+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results