accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2OOH

CAS RN 57718-07-7

IUPAC Name: 4-ethoxy-2-methylidene-4-oxobutanoic acid
Synonyms: 4-ethoxy-2-methylidene-4-oxobutanoic acid
Molecular Weight (g/mol): 158.15
Molecular Formula: C7H10O4
InChi Key: RTTAGBVNSDJDTE-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results