accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOH

CAS RN 5773-80-8

IUPAC Name: 6-bromonaphthalene-2-carboxylic acid
Synonyms: 6-bromonaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 251.08
Molecular Formula: C11H7BrO2
InChi Key: NPMCAVBMOTZUPD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=C(Br)C=CC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)