accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 57764-49-5

IUPAC Name: ethyl 1,2-benzoxazole-3-carboxylate
Synonyms: ethyl 1,2-benzoxazole-3-carboxylate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: IRTACBSCHHOIPA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NOC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results