accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OOH

CAS RN 57771-09-2

IUPAC Name: 4-(dibutylamino)-2-hydroxybenzaldehyde
Synonyms: 4-(dibutylamino)-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 249.35
Molecular Formula: C15H23NO2
InChi Key: BWHGDPKOFVNGFQ-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C1=CC(O)=C(C=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results