accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONCH3NH

CAS RN 577777-20-9

IUPAC Name: tert-butyl N-(azetidin-3-yl)-N-methylcarbamate
Synonyms: tert-butyl N-(azetidin-3-yl)-N-methylcarbamate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: LHUFQYUQIUJJIB-UHFFFAOYSA-N
SMILES: CN(C1CNC1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)