accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3O

CAS RN 5779-94-2

IUPAC Name: 2,5-dimethylbenzaldehyde
Synonyms: benzaldehyde, 2,5-dimethyl-
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: SMUVABOERCFKRW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(C=O)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)