accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 578-22-3

IUPAC Name: 5-hydroxy-2-methylbenzoic acid
Synonyms: 5-hydroxy-2-methylbenzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: ZIOYQUNKXJQXQY-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results