accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NNOO

CAS RN 578-46-1

IUPAC Name: 5-methyl-2-nitroaniline
Synonyms: 5-methyl-2-nitroaniline
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: IGDYNWKWXUCIJB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(N)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results