accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBr

CAS RN 578-57-4

IUPAC Name: 1-bromo-2-methoxybenzene
Synonyms: 1-bromo-2-methoxybenzene
Molecular Weight (g/mol): 187.04
Molecular Formula: C7H7BrO
InChi Key: HTDQSWDEWGSAMN-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1Br
Boiling Point 
  • (3)
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (8)