accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONH

CAS RN 578-95-0

IUPAC Name: 9,10-dihydroacridin-9-one
Synonyms: acridone 9(10H)-acridone 7H-dibenz(a,j)acrid-14-one 9(10h)-acridone acridanone 9-acridone
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: FZEYVTFCMJSGMP-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)