accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONBrI

CAS RN 578007-66-6

IUPAC Name: 5-bromo-3-iodo-2-methoxypyridine
Synonyms: 5-bromo-3-iodo-2-methoxypyridine
Molecular Weight (g/mol): 313.92
Molecular Formula: C6H5BrINO
InChi Key: ZKVDYKJRRHTPLT-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results