accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 578007-67-7

IUPAC Name: 2-amino-5-iodopyridine-3-carbaldehyde
Synonyms: 2-amino-5-iodopyridine-3-carbaldehyde
Molecular Weight (g/mol): 248.02
Molecular Formula: C6H5IN2O
InChi Key: DSXSLRYMXWHNPF-UHFFFAOYSA-N
SMILES: NC1=NC=C(I)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results